Sulfuric Acid

Supplier: ALDON CORP SE

SS1118-500ML SS1125-1L SS1125-500ML SS1126-500ML SS1118-2.5LR SS1120-500ML SS1121-1L SS1116-1L SS1127-1L SS1122-1L SS1126-1L SS1117-500ML SS1118B-PK/4R SS1115-2.5L SS1118-CONC SS1128-500ML AD-SS1118P-2.5L AD-SS1118P-PK/6
470045-604EA 41.7 CAD
470045-604 470302-856 470302-858 470302-868 470302-872 470302-852 470302-854 470302-860 470302-862 470302-864 470302-866 470302-870 470302-876 470302-878 470302-850 470225-682 470354-382 470354-388
Sulfuric Acid
Chemicals for Science Education

CAS Number: 7664-93-9
Formula: H2SO4
Density: 1.84 g/mL
Boiling and Freezing Point: 275-325°C, 11°C
Solubility: Water
Synonyms: Hydrogen Sulfate, Battery Acid
Shelf Life: 12 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.


Caution: This product is corrosive.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR