Sulfur

Supplier: ALDON CORP SE

SS1090-500G SS1090-100G SS1090-2.5KG SS1090-2.5KGP SS1085-100G SS1085-500G
470302-840EA 29.95 CAD
470302-840 470302-836 470302-838 470302-844 470302-842 470233-604
Sulfur
Chemicals for Science Education

CAS Number: 7704-34-9
Formula Weight: 32.06
Formula: S
Hazard Info: Flammable
Density (g/mL): 2.04-2.07
Boiling Point (°C): 444
Freezing Point (°C): 116-121
Solubility: Alcohol, Organic Solvents, Not Soluble in Water
Synonyms: Sulfur Flour
Shelf Life (months): 36
Storage: Red


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR