Sudan IV

Supplier: ALDON CORP SE

SS1070-100ML SS1070-500ML SS1070-100ML SS1060-10G
470302-826EA 23.3 CAD
470302-826 470302-828 470302-830 470302-832
Sudan IV
Chemicals for Science Education

CAS Number: 85-83-6
Formula: C24H20N4O
Boiling Point: 260°C
Solubility: Alcohol
Synonyms: Solvent Red 24, C.I. No. 26105
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR