Porcelain Crucibles with Covers
Supplier: American Scientific
5303-03 CH0076B 470006-206 J235 J233
470006-208EA 9.55 CAD
470006-208 470006-204 470006-206 470148-778 470149-028
Porcelain Crucibles with Covers
Crucibles
These crucibles are great for student use.
  • They withstand up to 1000 °C
  • Glazed except bottom and rim surfaces
  • Acid-resistant and chemically inert
  • Great for student use
  • Covers included
Except for the bottom and rim surfaces, the crucibles are glazed. They can withstand temperatures up to 1000 °C.
Each cover has a center-ring handle.