Student Wet Cell and Porous Cup
Supplier: VWR International
PH0927-N8 J267 PH0927PCUP
470017-192KT 82.25 CAD
470017-192 470148-894 470331-650
Student Wet Cell and Porous Cup
Educational Materials Chemistry Educational Materials Electrochemistry Learning Activities
Demonstrate Characteristics of Primary and Secondary Cells
Cover electrodes, electrolytes, circuits, electromotive force, polarization, depolarizers, and more with this complete outfit. The outfit includes a cell body of transparent, unbreakable plastic; a screw-on plastic ring; two brass electrode holders; 1 1/8" x 5" porous ceramic cup; ten electrodes: one each of aluminum, nickel, graphite, and iron; two each of copper, lead and zinc; and instructions.