Bacteria Experiment Kit
Supplier: CULTURE MEDIA &SUPPLIES INC SE
WL8163
470004-020KT 102 CAD
470004-020
Bacteria Experiment Kit
Educational Materials Biology Educational Materials Microbiology Learning Activities
Students find the answers to such questions as how much bacteria does soap or mouthwash eliminate, and which mouth contains more bacteria — a human or a dog.
  • Students conduct their own easy to perform, completely hands-on bacteria tests.
  • Collect and culture organisms from your environment
  • Comes with prepared media- just melt and pour
Students simply melt the provided media in a microwave, pour their own media plates, and test for growth using a series of experiments, including testing of common use items such as bathroom handles, door knobs and water fountains.
Ordering information: This item will ship immediately upon order, unless you specify otherwise.
Delivery information: This item ships directly from our supplier.