Ward's® Live Streptococcus pyogenes Culture - PATHOGEN
Supplier: Avantor
470179-210EA 22.05 CAD
470179-210 470179-570
Ward's® Live Streptococcus pyogenes Culture - PATHOGEN
Specimens Live Specimens Bacteria - Living Material
Lancefield group A, beta hemolysis. Sensitive to bacitracin. Causes strep throat, rheumatic fever, and scarlet fever.
  • Biosafety Level: 2
  • Canada: Containment Level 2 - PHAC Licence required
  • Motility: Negative
  • Morphology: Cocci in chains
  • Gram Reaction: Positive
  • Respiration: Aerobic but also grows well in 5% CO2
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
This organism is offered as a ready-to-use tube culture, as well as in a freeze-driedMore Product Information