Ward's® Live Streptococcus pneumoniae Culture - PATHOGEN
Supplier: Avantor
470179-172EA 22.05 CAD
470179-172 470179-498
Ward's® Live Streptococcus pneumoniae Culture - PATHOGEN
Specimens Live Specimens Bacteria - Living Material
No Lancefield group shown, alpha hemolysis. Bile solubility test positive, optochin sensitive. Causes pneumonia. 
  • Biosafety Level: 2
  • Canada: Containment Level 2 - PHAC Licence required
  • Motility: Negative
  • Morphology: Cocci in chains
  • Gram Reaction: Positive
  • Respiration: Aerobic
  • Hemolysis: Alpha
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
This organism is offered as a ready-to-use tube culture, as well as in a freeze-dried culture forMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable