Ward's® Live Streptococcus mutans Culture
Supplier: Avantor
470179-170EA 22.05 CAD
470179-170 470179-582
Ward's® Live Streptococcus mutans Culture
Specimens Live Specimens Bacteria - Living Material
Found on dental plaque, implicated in cavities. No Lancefield group shown, gamma hemolysis. 
  • Biosafety Level: 1
  • Motility: Negative
  • Morphology: Cocci in chains
  • Gram Reaction: Positive
  • Hemolysis: Gamma
  • Respiration: Aerobic
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
Ward’s Science is not affiliated with the American Type Culture Collection and does not provide ATCC numbers for our organisms.
Ordering information: Microbial and fungal cultures are restricted and can only be ordered by educational professionals.
These products contain living or perishable materials and will ship via overnight or second day delivery. Please specify a delivery date during checkout
Caution: While this organism is non-pathogenic, Ward’s recommends that you utilize proper aseptic technique when handling all microorganisms.