Glass Stirring Rods
Supplier: United Scientific Supplies
GSR006 GSR008 GSR005-PK/144 GSR012-W GSR015 GSR010
470020-786PK 16.25 CAD
470020-786 470020-788 470144-862 470189-016 470189-018 470200-955
Glass Stirring Rods
Stirrers Stirring Rods
Glass stirring rod features fire polished flint glass.
  • Rounded ends
Stirring rods are made of soft flint glass, with both ends fire-polished and rounded.