Ward's® Live Staphylococcus epidermidis Culture
Supplier: Avantor
470176-542EA 22.05 CAD
470176-542 470176-544
Ward's® Live Staphylococcus epidermidis Culture
Specimens Live Specimens Bacteria - Living Material
This organism is normal flora of human skin.
  • Biosafety Level: 1
  • Gram Stain: Positive
  • Morphology: Cocci in grape-like clusters
  • Coagulase: Negative
  • Catalase: Positive
  • Motility: Negative
  • Respiration: Aerobic
All bacterial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
This organism is offered as a ready-to-use tube culture, as well as in a freeze-dried culture for longer storage.
Freeze-dried organisms come with everything you need to rehydrate andMore Product Information