Boreal Standard Compound Microscopes
Supplier: VWR
SFC-111LED 5745000 5584400 5584123 5791102 5745200 5584135 5584500 MSH008
470135-366EA 448.8 CAD
470135-366 470145-560 470145-590 470145-716 470145-732 470145-714 470177-090 470177-148 470177-238 470014-588
Boreal Standard Compound Microscopes
Microscopes
Boreal standard microscopes feature metal construction, a rugged student-proof design, and precision optics making them a favorite among educators.
  • Coated-glass optics
  • Wide-field eyepiece with pointer
  • Limited lifetime warranty
These microscopes optics are parfocal and parcentered for a crisp, consistent field of view. All units feature strategically located controls, course and fine focus, and a 360° rotatable head. Units are available in a variety of configurations offering three or four objectives, plain or mechanical stage, and corded or cordlessMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Objectives 4×, 10×, 40× 4×, 10×, 40×, 100×