Microscope Slide Storage Cabinet
Supplier: Quincy Labs
100SCK-B
470039-314EA 617.4 CAD
470039-314
Microscope Slide Storage Cabinet
Slide Holders
Individual slide storage cabinets, which are stackable using interlocking tabs. Each cabinet has six drawers that include identification tabs for labeling and holds up to 5000 standard 3x1" glass slides vertically.
  • Keeps slides debris-free
  • Foam rubber dividers keep the slides upright when the drawers are only partially filled
  • Store slides neatly and efficiently
  • Construction prevents drawers from falling out accidentally
  • Made of heavy gauge steel with baked-on beige enamel finish, drawers have black phenolic knobs
Base size: 46.5x40x6.35 cm
Weight: 3.6 kg
Cabinet size: 46.5x37.4x12.7 cm
Weight: 14.5 kg