Squibb Urinometer and Jar
Supplier: Shiv Dial Sud & Sons
P11096 P11094 P11095
470148-982CS 1555.2 CAD
470148-982 470200-844 470200-952
Squibb Urinometer and Jar
Clinical Diagnostic Supplies Clinical Instruments
Perfect for when only minimal volumes are required, this urinometer provides highly accurate samples.
  • Calibrated, Easy-to-Read Scale
  • Range: 1.000–1.060 Sp. Gr
  • Accurate Samples
  • Stain-Free Clear Glass
  • Capacity: 50 mL
This stain-free and clear glass apparatus has a capacity of 50 mL and a wide base that prevents it from tipping.
Its range is 1.000–1.060 Sp. Gr in 0.002 graduations.