Speed Gun
Supplier: BUSHNELL CORPORATION
101911
470039-956EA 494.85 CAD
470039-956
Speed Gun
Educational Materials Physics Educational Materials Mechanics Learning Activities
Simple, Fast, and Fun
  • Easy point-and-shoot operation
  • A large, clear LCD display
Students are fascinated with speed. Unfortunately it is often tough to measure. This unit uses Bushnell Velocity to measure speed. This device is the ideal tool for tracking everything from racecars to pitching speeds — track the speed of a ball from 100 feet or a car up to 200 mph.
Baseball/softball/tennis: 6–110 mph
Auto racing 6–200 mph
Accuracy: ± 1.0 mph