CENCO® Electrodeless Spectrum Tube Power Supply
Supplier: Wards
SE1033-00 SE1034-00 SE1035-00 SE1037-00 SE1036-00 SE1030-00 SE1031-00 SE1032-00 SE1028-00
470013-412EA 99.55 CAD
470013-412 470013-414 470013-416 470013-420 470013-418 470013-422 470013-424 470017-046 470017-048
CENCO® Electrodeless Spectrum Tube Power Supply
Educational Materials Physics Educational Materials Optics Learning Activities
This safe spectrum power supply features encased spectrum tubes.
  • No Exposure to Voltage Source
  • No Electrodes in Bulbs
  • All Spectrum Tubes Are Encased Inside the Power Supply
  • For Use with Spectroscopes or Spectrometers
  • Bulbs Have 150 Hours of Burn Time
This innovative device far surpasses the safety, capability, and convenience of any other spectrum power supply available.
Most current power supplies expose students to 5000V. This device’s new, advanced design and features eliminate student’s contact to the voltage source. This safety feature allowsMore Product Information