Specific Heat Specimens Set
Supplier: VWR International
P20211
470148-828CS 3322.1 CAD
470148-828
Specific Heat Specimens Set
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
The Specific Heat Specimens Set can be used to determine the specific heats of a variety of common metals.
  • Five Cylinders Made of Copper, Tin, Zinc, and Aluminum
  • Each Cylinder is 1.3 dia.
  • For Determining Specific Heats of Common Metals
It consists of five cylinders of equal weight and made of lead, copper, tin, zinc, and aluminum. Each cylinder is 1.3 cm dia.
When brought to boiling water temperature and immersed in the same quantity of water at the same temperature, the rise in water temperature will be proportional to the specific heats.
Packaging: One package has five sets.