Live Sordaria fimicola Cultures
Supplier: AMERICAN TYPE CULTURE COLL.
AMCSORW AMCSORG AMCSORT AMCSORTD
470176-394EA 22.05 CAD
470176-394 470176-396 470176-398 470177-274 470177-284 470177-286 470179-718
Live Sordaria fimicola Cultures
Specimens Live Specimens Fungi - Living Material
Use Sordaria fimicola to allow students to visualize the results of crossing over during cell division.
  • Use living species to teach complex genetics concepts
  • High quality organisms cultured on-site and shipped to you ready to use
Sordaria's ascospores, exhibit characteristic color patterns based on mutant type and are readily observable under a microscope. Use the arrangement of ascospores to estimate percentage of crossover between the centromere and the gene that controls spore color.
Organisms can be crossed in any confiuration, but the mostMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable