Somso® Synapse Model
Supplier: MARCUS SOMMER
BS 35/3
470004-080EA 856.1 CAD
470004-080
Somso® Synapse Model
Models Human Biology Models
Nerve Components Enlarged 120,000X
  • Highly Detailed Section Of Axon Terminal
  • Shows Details Of Membrane Structure And Neurotransmitter Release
  • 5 Year Manufacturers Warranty
Based on the latest electron microscope interpretation, the model shows all the systems that play a part in the transmission of nerve impulse: neuro tubules and filaments, synaptic vesicles, and membrane structure. The model is mounted on a base and can be removed for closer examination.