Somso® Hominids Skull Restoration Series
Supplier: MARCUS SOMMER
S 1 S 2 S 3 S 5/1 S 3/1 S 5
470007-978EA 936.95 CAD
470007-978 470007-980 470007-982 470029-240 470029-242 470029-244 470029-246
Somso® Hominids Skull Restoration Series
Models Paleontology Models
Trace the Evolution of Humans with Accurate Reconstructions
  • Color Coded
  • Indicates parts that have been reconstructed
  • Removable lower jaw
Each skull is a durable, complete restoration cast in resin with a spring-held removable lower jaw. They are color coded with the dark areas indicating original fossils, and the lighter areas indicating reconstructed parts. The Proconsul africanus cast of a Miocene ape from Kenya, 1948, is dated at approximately 20 million years old. Australopithecus africanus is based on African finds 2 to 3 million years old. More Product Information