Somso® Dissectible Skulls
Supplier: MARCUS SOMMER
QS 8/2 QS 8/3
470030-012EA 1028.5 CAD
470030-012 470030-014
Somso® Dissectible Skulls
Models Human Biology Models
Fourteen Parts Show Relationship Between Sections Of Skull
  • Available In Natural , Or Didactic Colored Versions
  • Easy Peg Style Assembly
  • 5 Year Manufacturers Warranty
With these models, it’s easy for students to learn the shape, size, and relationships between the different parts of the skull as they take the life-size skull apart and put it back together. The plastic plugs on each piece make it quick and easy to fit each piece into the inserts. Removable parts include the sphenoid, occipital, and two temporal lobes; two parietal and one frontal lobe;More Product Information