Solar Demonstration Kit

Supplier: VWR International
PH50607A
470148-726CS 2701 CAD
470148-726
Solar Demonstration Kit
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
For Use With Solar Cell.

  • Contains interchangeable components
  • Design your own applications
  • Generate useable electricity

Each component in this kit connects to the solar cell. Expose the cell to light source and then demonstrate the effect of solar power in various devices. This kit includes a mounted 3 1/2" x 3 1/2" solar cell with terminals, a motor, a LED, a rechargeable battery, and an ammeter mounted on an 8" x 4 3/4" base.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR