Solar Cell Unit
Supplier: VWR International
PH1320
470016-014EA 100.35 CAD
470016-014
Solar Cell Unit
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Show How Direct/Indirect Sunlight Provides Power
Capable of running a small propeller, this solar cell comes mounted on a mini motor. Use this unit to demonstrate the conversion of solar energy into mechanical energy.