Soil Sampler Set
Supplier: OAKFIELD APPARATUS CO.
MODEL A
470001-822EA 739.89 CAD
470001-822
Soil Sampler Set
Samplers Water Samplers
Four-piece set of professional equipment.
  • Remove your core intact
  • Easy to use in the field
  • Aligned with GLOBE protocols
The 12" handle, auger, tube, and two 12" extension rods to reach a depth of 3', are all built to high-grade standards. The screw-on tube tip is replaceable to ensure lifetime usage of this handy tool. Weight: 4 lbs., 12 oz.