Soft Iron Rod
Supplier: Wards
S44370-35 S44370-37
470122-638PK 134.3 CAD
470122-638 470122-640
Soft Iron Rod
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Soft iron is a classic example of a readily magnetized material.
  • For Magnetic Demonstrations
  • Low-Remanence
  • Low-Coercivity
  • Easy-To-Magnetize
  • Ten Rods per Package
Since it has low remanence and coercivity, it is also readily demagnetized.
This soft iron rod is ideal for magnetic demonstrations.
Packaging: This item is available in packages of 10 pieces.
Order Now
SPECIFICATIONS
Environmentally Preferable