Sodium Thiosulfate, Anhydrous

Supplier: ALDON CORP SE

SS0882-500GR SS0882-100GR SS0900-500ML SS0899-500ML SS0894-500ML SS0897-500ML
470302-726EA 42.2 CAD
470302-726 470302-724 470302-728 470225-670 470225-810 470225-830
Sodium Thiosulfate, Anhydrous
Chemicals for Science Education

CAS Number: 7772-98-7
Formula Weight: 158.11
Formula: Na2S2O3
Density (g/mL): 1.667
Boiling Point (°C): >100
Freezing Point (°C): 48
Solubility: Water
Synonyms: Sodium Hyposulfite
Shelf Life (months): 12
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR