Sodium Thiocyanate

Supplier: ALDON CORP SE

SS0870-2.5KG SS0870-500G SS0869-500ML
470302-714EA 150.65 CAD
470302-714 470302-716 470225-748
Sodium Thiocyanate
Chemicals for Science Education

CAS Number: 540-72-7
Formula Weight: 81.07
Formula: NaSCN
Density (g/mL): 1.74
Freezing Point (°C): 287
Solubility: Water and Alcohol
Synonyms: Sodium Rhodante, Thiocyanic Acid, Sodium Salt
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR