Sodium Sulfite

Supplier: ALDON CORP SE

SS0850-500G SS0849-500GR SS0851-500ML
470302-710EA 27.65 CAD
470302-710 470302-712 470225-808
Sodium Sulfite
Chemicals for Science Education

CAS Number: 7757-83-7
Formula Weight: 126.04
Formula: Na2SO3
Hazard Info: Irritant
Density (g/mL): 1.4
Solubility: Water
Synonyms: Sulfurous Acid, Disodium Salt; Disodium Sulfite; Sodium Sulphite
Shelf Life (months): 12
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR