Sodium Sulfate

Supplier: ALDON CORP SE

SS0830-500G SS0841-50ML SS0830-500GRC SS0830-500GRG SS0830-2.5KG SS0830-500GP SS0841-500ML SS0842-500ML SS0828-500ML
470302-690EA 15.9 CAD
470302-690 470302-700 470302-688 470302-692 470302-694 470302-696 470302-702 470302-704 470225-672
Sodium Sulfate
Chemicals for Science Education

CAS Number: 7757-82-6
Formula Weight: 142.04
Formula: Na2SO4
Density (g/mL): 2.698
Boiling Point (°C): >1000
Freezing Point (°C): 888
Solubility: Water
Synonyms: Disodium Sulfate
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR