Sodium Silicate

Supplier: ALDON CORP SE

SS0820-1L SS0820-3.8L SS0820-500ML SS0805-500G
470302-676EA 36.8 CAD
470302-676 470302-678 470302-680 470302-682
Sodium Silicate
Chemicals for Science Education

CAS Number: 10213-79-3
Formula Weight: 212.03
Formula: Na2SiO3·5H2O
Density (g/mL): 1.81
Freezing Point (°C): 72.2
Synonyms: Disodium Trioxosilicate Pentahydrate
Shelf Life (months): 36
Storage: White


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR