Sodium Phosphate Tribasic, 12-Hydrate

Supplier: ALDON CORP SE

SS0790-500G SS0790-500GR SS0802-500ML
470302-670EA 36.5 CAD
470302-670 470302-668 470225-806
Sodium Phosphate Tribasic, 12-Hydrate
Chemicals for Science Education

CAS Number: 10101-89-0
Formula Weight: 380.12
Formula: Na3PO4·12H2O
Density (g/mL): 1.62
Freezing Point (°C): 73.3-76.7
Solubility: Water
Synonyms: Trisodium Phosphate, TSP
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR