Sodium Phosphate Monobasic Anhydrous

Supplier: ALDON CORP SE

SS0756-500G SS0756-500GR
470302-664EA 46.3 CAD
470302-664 470302-666
Sodium Phosphate Monobasic Anhydrous
Chemicals for Science Education

CAS Number: 7558-80-7
Formula Weight: 120.00
Formula: NaH2PO4
Density (g/mL): 2.047
Freezing Point (°C): 60
Synonyms: Sodium biphosphate, Sodium Dihydrogen Phosphate
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR