Sodium Oxalate

Supplier: ALDON CORP SE

SS0720-100GR SS0720-500GR SS0724-500ML
470302-652EA 116.7 CAD
470302-652 470302-654 470229-732
Sodium Oxalate
Chemicals for Science Education

CAS Number: 62-76-0
Formula Weight: 134.00
Formula: (COONa)2
Hazard Info: Toxic
Density (g/mL): 2.34
Freezing Point (°C): 250-270
Synonyms: Ethanedioic Acid Disodium Salt
Shelf Life (months): 12
Storage: White


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR