Sodium Nitrate

Supplier: ALDON CORP SE

SS0690-500G SS0691-500ML SS0693-500ML SS0690-2.5KG SS0680-100GR SS0680-500GR SS0690-100G SS0690-2.5KGP SS0692-CONC SS0692-500ML
470302-640EA 29.8 CAD
470302-640 470302-644 470302-646 470302-630 470302-632 470302-634 470302-638 470302-642 470302-648 470225-740
Sodium Nitrate
Chemicals for Science Education

CAS Number: 7631-99-4
Formula Weight: 84.99
Formula: NaNO3
Hazard Info: Oxidizer, Explosive, Toxic
Density (g/mL): 2.261
Freezing Point (°C): 307
Solubility: Water and Alcohol
Synonyms: Nitrate of Soda, Chile Saltpeter
Shelf Life (months): 36
Storage: Yellow


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR