Sodium Iodate

Supplier: ALDON CORP SE

SS0615-25G
470302-592EA 56.4 CAD
470302-592
Sodium Iodate
Chemicals for Science Education

CAS Number: 7681-55-2
Formula Weight: 197.90
Formula: NaIO3
Density (g/mL): 4.277
Solubility: Water, Acetone
Synonyms: Iodic Acid Sodium Salt
Shelf Life (months): 12
Storage: Yellow


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR