Sodium Hypochlorite

Supplier: ALDON CORP SE

SS0610-3.8L SS0610-1L SS0610-500ML
470302-588EA 48.1 CAD
470302-588 470302-586 470302-590
Sodium Hypochlorite
Chemicals for Science Education

CAS Number: 7681-52-9
Formula Weight: 1.2
Formula: NaOCl
Solubility: Water
Synonyms: Chlorine Bleach, Bleach
Shelf Life (months): 24
Storage: White


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR