Sodium Hydroxide

Supplier: ALDON CORP SE

SS0550-2.5KG SS0550-100GR SS0550-2.5KGR SS0600-1L SS0600-500ML SS0582-500ML SS0580-1L SS0580-500ML SS0580-3.8L SS0575-1L SS0576-500ML SS0550-500G SS0570-CONC SS0570-2.5KG SS0570-500G SS0550-500GR SS0600-3.8L SS0582-1L SS0595-500ML SS0574-500ML SS0581-500ML SS0593-100ML SS0601-500ML
470302-548EA 11.85 CAD
470302-548 470302-550 470302-554 470302-556 470302-560 470302-564 470302-568 470302-570 470302-572 470302-574 470302-578 470302-582 470302-552 470302-542 470302-544 470302-546 470302-558 470302-562 470302-566 470302-576 470302-580 470225-676 470225-802 470225-804
Sodium Hydroxide
Chemicals for Science Education

CAS Number: 1310-73-2
Formula: NaOH
Density: 2.13 g/mL
Boiling and Freezing Point: 1390°C, 318°C
Solubility: Water and Alcohol
Synonyms: Caustic Soda
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.


Caution: This product is corrosive and toxic.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR