Sodium Fluoride

Supplier: ALDON CORP SE

SS0534-50ML SS0535-500G SS0535-100GR SS0534-500ML
470302-536EA 21.25 CAD
470302-536 470302-538 470302-540 470225-800
Sodium Fluoride
Chemicals for Science Education

CAS Number: 7681-49-4
Formula Weight: 41.99
Formula: NaF
Hazard Info: Toxic
Density (g/mL): 2.78
Boiling Point (°C): 1704
Freezing Point (°C): 993
Solubility: Water and Alcohol
Synonyms: Fluoridine
Shelf Life (months): 36
Storage: Blue


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR