Sodium Dodecyl Sulfate

Supplier: ALDON CORP SE

SS0647-100ML SS0648-50ML SS0650-100G SS0650-500G SS0650-25GR
470302-610EA 17.05 CAD
470302-610 470302-612 470302-614 470302-616 470302-618
Sodium Dodecyl Sulfate
Chemicals for Science Education

CAS Number: 151-21-3
Formula: CH3CH211OSO3Na
Freezing Point: 204 to 207°C
Solubility: Water and Alcohol
Synonyms: Sodium Lauryl Sulfate, SDS
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR