Sodium Chromate Anhydrous
Supplier: ALDON CORP SE
CAS Number: 7775-11-3
Formula Weight: 161.99
Formula: Na2CrO4
Hazard Info: Toxic
Density (g/mL): 2.723
Freezing Point (°C): 792
Solubility: Water and Alcohol
Synonyms: Disodium Chromate(VI), Chromate of Soda
Shelf Life (months): 36
Storage: Blue
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...