Sodium Chloride

Supplier: ALDON CORP SE

SS0450-2.5KG SS0430-2.5KG SS0430-2.5KGR SS0442-50ML SS0430-CAPS SS0450-500G SS0442-500ML SS0441-500ML SS0430-12KG SS0430-735G SS0430-500GR SS0440-1L SS0436-500ML SS0437-1L
470302-498EA 22.45 CAD
470302-498 470302-512 470302-520 470302-502 470302-494 470302-500 470302-504 470302-506 470302-510 470302-516 470302-522 470225-730 470225-796 470225-798
Sodium Chloride
Chemicals for Science Education

CAS Number: 7647-14-5
Formula: NaCl
Density: 2.16 g/mL
Boiling and Freezing Point: 1465°C, 801°C
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR