Sodium Chlorate

Supplier: ALDON CORP SE

SS0410-100G SS0410-500G SS0410-500GR
470302-488EA 46.7 CAD
470302-488 470302-490 470302-492
Sodium Chlorate
Chemicals for Science Education

CAS Number: 7775-09-9
Formula Weight: 106.44
Formula: NaClO3
Hazard Info: Oxidizer, Flammable
Density (g/mL): 2.490
Freezing Point (°C): 248-261
Solubility: Water and Alcohol
Shelf Life (months): 36
Storage: Yellow


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR