Sodium Carbonate Monohydrate

Supplier: ALDON CORP SE

SS0380-500G SS0403-1L SS0403-50ML
470302-478EA 48.75 CAD
470302-478 470302-482 470302-486
Sodium Carbonate Monohydrate
Chemicals for Science Education

CAS Number: 5968-11-6
Formula Weight: 124.00
Formula: Na2CO3·H2O
Hazard Info: Irritant
Density (g/mL): 2.25
Freezing Point (°C): 100
Solubility: Water and Glycerin
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR