Sodium Carbonate

Supplier: ALDON CORP SE

SS0360-2.5KG SS0360-500GR SS0403-500ML SS0360-CAPS SS0402-CONC SS0402-500ML SS0408-500ML
470302-468EA 44.3 CAD
470302-468 470302-476 470302-480 470302-472 470302-474 470302-484 470225-688
Sodium Carbonate
Chemicals for Science Education

CAS Number: 497-19-8
Formula: Na2CO3
Density: 2.533 g/mL
Freezing Point: 864°C
Solubility: Water and Glycerin
Synonyms: Soda Ash
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.


Caution: This product is an irritant.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR