Sodium Bromide

Supplier: ALDON CORP SE

SS0349-500ML SS0340-100G SS0340-500G SS0340-100GR SS0340-500GR SS0351-500ML SS0352-1L SS0353-500ML
470302-458EA 25.5 CAD
470302-458 470302-460 470302-462 470302-464 470302-466 470225-708 470225-732 470225-746
Sodium Bromide
Chemicals for Science Education

CAS Number: 7647-15-6
Formula Weight: 102.90
Formula: NaBr
Hazard Info: Toxic
Density (g/mL): 3.203
Boiling Point (°C): 1391
Freezing Point (°C): 755
Solubility: Water and Alcohol
Synonyms: Sedoneual
Shelf Life (months): 12
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR