Sodium Borate, 10-Hydrate

Supplier: ALDON CORP SE

SS0320-500G SS0320-500GR SS0320-100GR
470302-452EA 24.9 CAD
470302-452 470302-454 470302-456 470233-596
Sodium Borate, 10-Hydrate
Chemicals for Science Education

CAS Number: 1303-96-4
Formula: Na2B4O7 10H2O
Density: 1.73 g/mL
Freezing Point: 62 °C
Solubility: Water and Glycerin
Synonyms: Sodium Tetraborate, Borax
Shelf Life: 12 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR