Sodium Bisulfite

Supplier: ALDON CORP SE

SS0305-500G SS0305-500GR
470302-448EA 28.2 CAD
470302-448 470302-450
Sodium Bisulfite
Chemicals for Science Education

CAS Number: 7631-90-5
Formula Weight: 104.07
Formula: NaHSO3
Density (g/mL): 1.48
Freezing Point (°C): 150
Solubility: Water
Synonyms: Sodium Hydrogen Sulfite
Shelf Life (months): 12
Storage: White


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR