Sodium Acetate Trihydrate

Supplier: ALDON CORP SE

SS0232-CONC SS0233-100GR SS0233-500GR
470302-426EA 18.9 CAD
470302-426 470302-428 470302-430
Sodium Acetate Trihydrate
Chemicals for Science Education

CAS Number: 6131-90-4
Formula: NaC2H3O2 3H2O
Density: 1.45 g/mL
Boiling and Freezing Point: 122.7°C, 57.7°C
Shelf Life: 12 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR