Sodium

Supplier: ALDON CORP SE

SS0215-10G SS0216-100G SS0217-100GR SS0215-1G SS0215-1.5G
470302-622EA 124.2 CAD
470302-622 470302-624 470302-626 470302-732 470302-628
Sodium
Chemicals for Science Education

CAS Number: 7440-23-5
Formula: Na
Density: 0.97 g/mL
Boiling and Freezing Point: 881°C, 98°C
Synonyms: Natrium
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.


Caution: This product is flammable and dangerous when wet.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR