Ward's® Slate (Red)
Supplier: Avantor
470026-276PK 21.3 CAD
470026-276 470026-278 470026-280
Ward's® Slate (Red)
Specimens Rock Specimens Metamorphic Rocks
Slaty cleavage; contains hematite.
Ordering information: WARD’S offers different specimen sizes to meet your various needs. Student specimens are ideal for students to use for study and identification. Student specimens range in size from 1×1" to 1×2" and come in packages of ten. Hand specimens are great for lectures, demonstrations, and display. Hand specimens range in size from 2×3" to 3×4", and each comes in a labeled plastic tray. Bulk pack includes several of the smaller pieces that are suitable for classroom study, while the larger specimens in the one kilogram pack can be cut toMore Product Information